missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dimethylnaphthalene 98.0+%, TCI America™

Catalog No. p-7126411
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D07521G 1 g
D07525G 5 g
2 options

Catalog No. D07521G

Supplier: TCI America D07521G

Only null left

Chemical Identifiers

CAS 582-16-1
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
MDL Number MFCD00004121
InChI Key LRQYSMQNJLZKPS-UHFFFAOYSA-N
Synonym naphthalene, 2,7-dimethyl, 2,7-dmn, unii-on63xh5bq4, 2,7-dimethyl-naphthalene, on63xh5bq4, acmc-1ar8i, 2,7-dimethylnaphthalene, naphthalene, 2,7-dimethyl-8ci 9ci
PubChem CID 11396
ChEBI CHEBI:48632
IUPAC Name 2,7-dimethylnaphthalene
SMILES CC1=CC2=CC(C)=CC=C2C=C1

Specifications

Melting Point 97°C
Color White-Yellow
Boiling Point 263°C
Quantity 1 g
Formula Weight 156.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dimethylnaphthalene