missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dimethylnaphthalene, 99%

Catalog No. AAB2170403
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2170403 1 g
AAB2170401 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2170403 Supplier Thermo Scientific Chemicals Supplier No. B2170403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 582-16-1 | C12H12 | 156.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 582-16-1
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
MDL Number MFCD00004121
InChI Key LRQYSMQNJLZKPS-UHFFFAOYSA-N
Synonym naphthalene, 2,7-dimethyl, 2,7-dmn, unii-on63xh5bq4, 2,7-dimethyl-naphthalene, on63xh5bq4, acmc-1ar8i, 2,7-dimethylnaphthalene, naphthalene, 2,7-dimethyl-8ci 9ci
PubChem CID 11396
ChEBI CHEBI:48632
IUPAC Name 2,7-dimethylnaphthalene
SMILES CC1=CC2=CC(C)=CC=C2C=C1

Specifications

Melting Point 96°C to 98°C
Boiling Point 262°C to 264°C
Quantity 1 g
Beilstein 1852737
Formula Weight 156.23
Percent Purity 99%
Chemical Name or Material 2,7-Dimethylnaphthalene
EINECSNumber 209-477-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only