missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dimethoxy-9H-carbazole 98.0+%, TCI America™
$251.65 - $753.08
Chemical Identifiers
| CAS | 61822-18-2 |
|---|---|
| Molecular Formula | C14H13NO2 |
| Molecular Weight (g/mol) | 227.26 |
| MDL Number | MFCD28154716 |
| InChI Key | OFGBQGFYHXYVIA-UHFFFAOYSA-N |
| PubChem CID | 12339399 |
| IUPAC Name | 2,7-dimethoxy-9H-carbazole |
| SMILES | COC1=CC2=C(C=C1)C1=C(N2)C=C(OC)C=C1 |
Chemical Identifiers
| 61822-18-2 | |
| 227.26 | |
| OFGBQGFYHXYVIA-UHFFFAOYSA-N | |
| 2,7-dimethoxy-9H-carbazole |
| C14H13NO2 | |
| MFCD28154716 | |
| 12339399 | |
| COC1=CC2=C(C=C1)C1=C(N2)C=C(OC)C=C1 |
Specifications
| 61822-18-2 | |
| C14H13NO2 | |
| 1 g | |
| COC1=CC2=C(C=C1)C1=C(N2)C=C(OC)C=C1 | |
| 227.26 | |
| 227.26 | |
| Crystalline Powder |
| 282°C | |
| MFCD28154716 | |
| OFGBQGFYHXYVIA-UHFFFAOYSA-N | |
| 2,7-dimethoxy-9H-carbazole | |
| 12339399 | |
| ≥98.0% (GC) | |
| 2,7-Dimethoxy-9H-carbazole |
Safety and Handling
TSCA : No