missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dimethoxy-9H-carbazole 98.0+%, TCI America™

Catalog No. D53295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53295G 5 g
D53291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53295G Supplier TCI America Supplier No. D53295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61822-18-2
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD28154716
InChI Key OFGBQGFYHXYVIA-UHFFFAOYSA-N
PubChem CID 12339399
IUPAC Name 2,7-dimethoxy-9H-carbazole
SMILES COC1=CC2=C(C=C1)C1=C(N2)C=C(OC)C=C1

Specifications

Melting Point 282°C
Quantity 5 g
Formula Weight 227.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dimethoxy-9H-carbazole