missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dicyanonaphthalene 94.0+%, TCI America™
Specifications
Specifications
| Melting Point | 267°C |
| Color | Yellow |
| Quantity | 1 g |
| UN Number | 3439 |
| Formula Weight | 178.19 |
| Percent Purity | ≥94.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,7-Dicyanonaphthalene |
Safety and Handling
| TSCA | No |