missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ 2',7'-Dichlorofluorescin Diacetate, Calbiochem™,

Catalog No. 287810100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
287810100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 287810100MG Supplier MilliporeSigma™ Supplier No. 287810100MG

Cell-permeable fluorogenic probe that is useful for the detection of reactive oxygen species (ROS) and nitric oxide (·NO) and for the determination of the degree of overall oxidative stress

Chemical Identifiers

CAS 4091-99-0
Molecular Formula C24H16Cl2O7
Molecular Weight (g/mol) 487.285
InChI Key PXEZTIWVRVSYOK-UHFFFAOYSA-N
Synonym h2dcfda, 2-3,6-diacetoxy-2,7-dichloro-9h-xanthen-9-yl benzoic acid, 2',7'-dichlorohydrofluorescein diacetate, 2,7-dichlorodihydrofluorescein diacetate, benzoic acid, 2-3,6-bis acetyloxy-2,7-dichloro-9h-xanthen-9-yl, 2-3,6-bis acetyloxy-2,7-dichloro-9h-xanthen-9-yl benzoic acid, 2′,7′-dichlorofluorescin diacetate, 2′,7′-dichlorodihydrofluorescein diacetate, 2-3,6-diacetyloxy-2,7-dichloro-9h-xanthen-9-yl benzoic acid
PubChem CID 77718
IUPAC Name 2-(3,6-diacetyloxy-2,7-dichloro-9H-xanthen-9-yl)benzoic acid
SMILES CC(=O)OC1=C(C=C2C(C3=CC(=C(C=C3OC2=C1)OC(=O)C)Cl)C4=CC=CC=C4C(=O)O)Cl

Specifications

Quantity 100 mg
Solubility Information Soluble in DSMO
Formula Weight 487.3
Percent Purity ≥94%
Grade HPLC
Physical Form Crystalline Solid
Chemical Name or Material 2',7'-Dichlorofluorescin Diacetate
Recommended Storage -20°C (-4°F)