missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromonaphthalene 98.0+%, TCI America™

$283.00 - $1062.69

Chemical Identifiers

CAS 58556-75-5
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD00238580
InChI Key ODJZWBLNJKNOJK-UHFFFAOYSA-N
Synonym naphthalene, 2,7-dibromo, acmc-209m5e, ksc490o3j, 2,7-dibromonaphthalene hplc
PubChem CID 625355
IUPAC Name 2,7-dibromonaphthalene
SMILES BrC1=CC2=CC(Br)=CC=C2C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D36245G
View Documents
TCI America
D36245G
5 g
Each for $283.00
Only null left
 
D362425G
View Documents
TCI America
D362425G
25 g
Each for $1,062.69
Only null left
 
Description

Description

Specifications

Chemical Identifiers

58556-75-5
285.97
ODJZWBLNJKNOJK-UHFFFAOYSA-N
625355
BrC1=CC2=CC(Br)=CC=C2C=C1
C10H6Br2
MFCD00238580
naphthalene, 2,7-dibromo, acmc-209m5e, ksc490o3j, 2,7-dibromonaphthalene hplc
2,7-dibromonaphthalene

Specifications

58556-75-5
White-Yellow
MFCD00238580
naphthalene, 2,7-dibromo, acmc-209m5e, ksc490o3j, 2,7-dibromonaphthalene hplc
BrC1=CC2=CC(Br)=CC=C2C=C1
285.97
285.97
Crystalline Powder
141°C
C10H6Br2
5 g
ODJZWBLNJKNOJK-UHFFFAOYSA-N
2,7-dibromonaphthalene
625355
≥98.0% (GC)
2,7-Dibromonaphthalene
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents