missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromocarbazole

Catalog No. AC467780010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467780010 1 g
AC467780050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467780010 Supplier Thermo Scientific Chemicals Supplier No. 467780010

CAS: 136630-39-2 | C12H7Br2N | 325.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136630-39-2
Molecular Formula C12H7Br2N
Molecular Weight (g/mol) 325.00
InChI Key QPTWWBLGJZWRAV-UHFFFAOYSA-N
IUPAC Name 2,7-dibromo-9H-carbazole
SMILES BrC1=CC2=C(C=C1)C1=C(N2)C=C(Br)C=C1

Specifications

Melting Point 225.0°C to 230.0°C
Quantity 1 g
Formula Weight 325
Chemical Name or Material 2,7-Dibromocarbazole
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement Toxic if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

missing translation for 'healthHazard3' GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only