missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromocarbazole 98.0+%, TCI America™

Catalog No. D39321G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39321G 1 g
D39325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39321G Supplier TCI America Supplier No. D39321G

Chemical Identifiers

CAS 136630-39-2
Molecular Formula C12H7Br2N
Molecular Weight (g/mol) 325.003
MDL Number MFCD09033507
InChI Key QPTWWBLGJZWRAV-UHFFFAOYSA-N
PubChem CID 11151503
IUPAC Name 2,7-dibromo-9H-carbazole
SMILES C1=CC2=C(C=C1Br)NC3=C2C=CC(=C3)Br

Specifications

Melting Point 232°C
Color White-Yellow
Quantity 1 g
Formula Weight 325.00
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromocarbazole