missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9-hexylcarbazole 98.0+%, TCI America™

Catalog No. D50551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50551G 1 g
D50555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50551G Supplier TCI America Supplier No. D50551G
Only null left

Chemical Identifiers

CAS 654676-12-7
Molecular Formula C18H19Br2N
Molecular Weight (g/mol) 409.165
InChI Key YSUUFISKPJBPOH-UHFFFAOYSA-N
PubChem CID 25150458
IUPAC Name 2,7-dibromo-9-hexylcarbazole
SMILES CCCCCCN1C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 1 g
Formula Weight 409.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-hexylcarbazole