missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dibromo-9-heptyl-9H-carbazole 98.0+%, TCI America™
$277.75 - $825.78
Chemical Identifiers
| CAS | 1173071-58-3 |
|---|---|
| Molecular Formula | C19H21Br2N |
| Molecular Weight (g/mol) | 423.192 |
| InChI Key | MLLYLIXYBBORGG-UHFFFAOYSA-N |
| PubChem CID | 129319314 |
| IUPAC Name | 2,7-dibromo-9-heptylcarbazole |
| SMILES | CCCCCCCN1C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br |
Chemical Identifiers
| 1173071-58-3 | |
| 423.192 | |
| 129319314 | |
| CCCCCCCN1C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br |
| C19H21Br2N | |
| MLLYLIXYBBORGG-UHFFFAOYSA-N | |
| 2,7-dibromo-9-heptylcarbazole |
Specifications
| 1173071-58-3 | |
| White | |
| 1 g | |
| CCCCCCCN1C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br | |
| 423.192 | |
| 423.19 | |
| Crystalline Powder |
| 74°C | |
| C19H21Br2N | |
| MLLYLIXYBBORGG-UHFFFAOYSA-N | |
| 2,7-dibromo-9-heptylcarbazole | |
| 129319314 | |
| ≥98.0% (GC) | |
| 2,7-Dibromo-9-heptyl-9H-carbazole |
Safety and Handling
TSCA : No