missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9-(2-ethylhexyl)carbazole 98.0+%, TCI America™

Catalog No. D4735200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4735200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4735200MG Supplier TCI America Supplier No. D4735200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 544436-46-6
Molecular Formula C20H23Br2N
Molecular Weight (g/mol) 437.219
MDL Number MFCD23703116
InChI Key OLOMDFPWMJQODN-UHFFFAOYSA-N
PubChem CID 12070511
IUPAC Name 2,7-dibromo-9-(2-ethylhexyl)carbazole
SMILES CCCCC(CC)CN1C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 200 mg
Formula Weight 437.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-(2-ethylhexyl)carbazole