missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Di-tert-butylpyrene 98.0+%, TCI America™
$136.69 - $411.98
Chemical Identifiers
| CAS | 24300-91-2 |
|---|---|
| Molecular Formula | C24H26 |
| Molecular Weight (g/mol) | 314.47 |
| MDL Number | MFCD01013641 |
| InChI Key | SKZSSCAGJZEXEA-UHFFFAOYSA-N |
| PubChem CID | 185563 |
| IUPAC Name | 2,7-di-tert-butylpyrene |
| SMILES | CC(C)(C)C1=CC2=CC=C3C=C(C=C4C=CC(=C1)C2=C34)C(C)(C)C |
Chemical Identifiers
| 24300-91-2 | |
| 314.47 | |
| SKZSSCAGJZEXEA-UHFFFAOYSA-N | |
| 2,7-di-tert-butylpyrene |
| C24H26 | |
| MFCD01013641 | |
| 185563 | |
| CC(C)(C)C1=CC2=CC=C3C=C(C=C4C=CC(=C1)C2=C34)C(C)(C)C |
Specifications
| 24300-91-2 | |
| White-Yellow | |
| MFCD01013641 | |
| SKZSSCAGJZEXEA-UHFFFAOYSA-N | |
| 2,7-di-tert-butylpyrene | |
| 185563 | |
| ≥98.0% (GC) | |
| 2,7-Di-tert-butylpyrene |
| 209°C | |
| C24H26 | |
| 1 g | |
| CC(C)(C)C1=CC2=CC=C3C=C(C=C4C=CC(=C1)C2=C34)C(C)(C)C | |
| 314.47 | |
| 314.47 | |
| Crystalline Powder |
Safety and Handling
TSCA : No