missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Bis[N-(m-tolyl)anilino]-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™
$240.46 - $829.51
Chemical Identifiers
| CAS | 1033035-83-4 |
|---|---|
| Molecular Formula | C51H38N2 |
| Molecular Weight (g/mol) | 678.879 |
| MDL Number | MFCD11112143 |
| InChI Key | QZTQQBIGSZWRGI-UHFFFAOYSA-N |
| Synonym | 2,7-Bis[phenyl(m-tolyl)amino]-9,9′C-spirobi[9H-fluorene] |
| PubChem CID | 51358300 |
| IUPAC Name | 2-N',7-N'-bis(3-methylphenyl)-2-N',7-N'-diphenyl-9,9'-spirobi[fluorene]-2',7'-diamine |
| SMILES | CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=CC4=C(C=C3)C5=C(C46C7=CC=CC=C7C8=CC=CC=C68)C=C(C=C5)N(C9=CC=CC=C9)C1=CC=CC(=C1)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4882200MG
|
TCI America
B4882200MG |
200 mg |
Each for $240.46
|
|
|||||
|
B48821G
|
TCI America
B48821G |
1 g |
Each for $829.51
|
|
|||||
Chemical Identifiers
| 1033035-83-4 | |
| 678.879 | |
| QZTQQBIGSZWRGI-UHFFFAOYSA-N | |
| 51358300 | |
| CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=CC4=C(C=C3)C5=C(C46C7=CC=CC=C7C8=CC=CC=C68)C=C(C=C5)N(C9=CC=CC=C9)C1=CC=CC(=C1)C |
| C51H38N2 | |
| MFCD11112143 | |
| 2,7-Bis[phenyl(m-tolyl)amino]-9,9′C-spirobi[9H-fluorene] | |
| 2-N',7-N'-bis(3-methylphenyl)-2-N',7-N'-diphenyl-9,9'-spirobi[fluorene]-2',7'-diamine |
Specifications
| 1033035-83-4 | |
| Yellow | |
| MFCD11112143 | |
| 2,7-Bis[phenyl(m-tolyl)amino]-9,9′C-spirobi[9H-fluorene] | |
| CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=CC4=C(C=C3)C5=C(C46C7=CC=CC=C7C8=CC=CC=C68)C=C(C=C5)N(C9=CC=CC=C9)C1=CC=CC(=C1)C | |
| 678.879 | |
| 678.88 | |
| Crystalline Powder |
| 213°C | |
| C51H38N2 | |
| 200 mg | |
| QZTQQBIGSZWRGI-UHFFFAOYSA-N | |
| 2-N',7-N'-bis(3-methylphenyl)-2-N',7-N'-diphenyl-9,9'-spirobi[fluorene]-2',7'-diamine | |
| 51358300 | |
| ≥98.0% (HPLC,N) | |
| 2,7-Bis[N-(m-tolyl)anilino]-9,9′-spirobi[9H-fluorene] |
Safety and Handling
TSCA : No