missing translation for 'onlineSavingsMsg'
Learn More

2,7-Bis[N-(1-naphthyl)anilino]-9,9-dimethylfluorene 98.0+%, TCI America™

Catalog No. B4926200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4926200MG 200 mg
B49261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4926200MG Supplier TCI America Supplier No. B4926200MG

Chemical Identifiers

CAS 222319-05-3
Molecular Formula C47H36N2
Molecular Weight (g/mol) 628.82
MDL Number MFCD11110702
InChI Key KJEQVQJWXVHKGT-UHFFFAOYSA-N
Synonym 9,9-Dimethyl-N,N′C-di(1-naphthyl)-N,N′C-diphenyl-9H-fluorene-2,7-diamine, DMFL-NPB
PubChem CID 20745597
IUPAC Name 9,9-dimethyl-N2,N7-bis(naphthalen-1-yl)-N2,N7-diphenyl-9H-fluorene-2,7-diamine
SMILES CC1(C)C2=C(C=CC(=C2)N(C2=CC=CC=C2)C2=C3C=CC=CC3=CC=C2)C2=C1C=C(C=C2)N(C1=CC=CC=C1)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 267°C
Color White-Yellow
Quantity 200 mg
Formula Weight 628.82
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Bis[N-(1-naphthyl)anilino]-9,9-dimethylfluorene