missing translation for 'onlineSavingsMsg'
Learn More

2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene 97.0+%, TCI America™

Catalog No. B39561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39561G 1 g
B39565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39561G Supplier TCI America Supplier No. B39561G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 688756-58-3
Molecular Formula C28H32B2O4
MDL Number MFCD15143601
Synonym Pyrene-2,7-diboronic Acid Bis(pinacol) Ester

Specifications

Color White
Quantity 1 g
Formula Weight 454.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene