missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedimethanol 98.0+%, TCI America™

Catalog No. p-7134775
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08035G 5 g
P080325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08035G Supplier TCI America Supplier No. P08035G
Only null left

Chemical Identifiers

CAS 1195-59-1
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00006351
InChI Key WWFMINHWJYHXHF-UHFFFAOYSA-N
Synonym 2,6-pyridinedimethanol, pyridine-2,6-diyldimethanol, 2,6-bis hydroxymethyl pyridine, 6-hydroxymethyl pyridin-2-yl methanol, pyridine-2,6-dimethanol, 2,6-dihydroxymethylpyridine, 2,6-bis-hydroxymethyl pyridine, 2,6-bis-hydroxymethyl-pyridine, 2,6-pyridinyl dimethanol, 6-hydroxymethyl-2-pyridyl methan-1-ol
PubChem CID 70957
IUPAC Name [6-(hydroxymethyl)pyridin-2-yl]methanol
SMILES C1=CC(=NC(=C1)CO)CO

Specifications

Melting Point 113°C
Color White
Boiling Point 185°C
Quantity 5 g
Formula Weight 139.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Pyridinedimethanol