missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedimethanol, 98%

Catalog No. AC167550050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167550050 5 g Glass bottle
AC167550250 25 g Glass bottle
AC167551000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC167550050 Supplier Thermo Scientific Chemicals Supplier No. 167550050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1195-59-1 | C7H9NO2 | 139.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1195-59-1
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.15
MDL Number MFCD00006351
InChI Key WWFMINHWJYHXHF-UHFFFAOYSA-N
Synonym 2,6-pyridinedimethanol, pyridine-2,6-diyldimethanol, 2,6-bis hydroxymethyl pyridine, 6-hydroxymethyl pyridin-2-yl methanol, pyridine-2,6-dimethanol, 2,6-dihydroxymethylpyridine, 2,6-bis-hydroxymethyl pyridine, 2,6-bis-hydroxymethyl-pyridine, 2,6-pyridinyl dimethanol, 6-hydroxymethyl-2-pyridyl methan-1-ol
PubChem CID 70957
IUPAC Name [6-(hydroxymethyl)pyridin-2-yl]methanol
SMILES C1=CC(=NC(=C1)CO)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 112°C to 115°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)(CH2OH)2
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 139.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Pyridinedimethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 214-803-

RUO – Research Use Only