missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedimethanol, 98%

CAS: 1195-59-1 | C7H9NO2 | 139.154 g/mol

Supplier:  Thermo Scientific Chemicals L0252714

 View more versions of this product

Catalog No. AAL0252714


Only null left
Explore available promotions

Description

Description

2,6-Pyridinedimethanol was used in the preparation of Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) and Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)). It is a ligand used in the synthesis of a variety of metal complexes and catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Pyridinedimethanol was used in the preparation of Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) and Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)). It is a ligand used in the synthesis of a variety of metal complexes and catalysts.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

2,6-Pyridinedimethanol
110°C to 114°C
C7H9NO2
25 g
2,6-pyridinedimethanol, pyridine-2,6-diyldimethanol, 2,6-bis hydroxymethyl pyridine, 6-hydroxymethyl pyridin-2-yl methanol, pyridine-2,6-dimethanol, 2,6-dihydroxymethylpyridine, 2,6-bis-hydroxymethyl pyridine, 2,6-bis-hydroxymethyl-pyridine, 2,6-pyridinyl dimethanol, 6-hydroxymethyl-2-pyridyl methan-1-ol
WWFMINHWJYHXHF-UHFFFAOYSA-N
[6-(hydroxymethyl)pyridin-2-yl]methanol
70957
98%
1195-59-1
185°C (15 mmHg)
MFCD00006351
116016
Soluble in water.
C1=CC(=NC(=C1)CO)CO
139.154
139.15

Chemical Identifiers

1195-59-1
139.154
WWFMINHWJYHXHF-UHFFFAOYSA-N
70957
C1=CC(=NC(=C1)CO)CO
C7H9NO2
MFCD00006351
2,6-pyridinedimethanol, pyridine-2,6-diyldimethanol, 2,6-bis hydroxymethyl pyridine, 6-hydroxymethyl pyridin-2-yl methanol, pyridine-2,6-dimethanol, 2,6-dihydroxymethylpyridine, 2,6-bis-hydroxymethyl pyridine, 2,6-bis-hydroxymethyl-pyridine, 2,6-pyridinyl dimethanol, 6-hydroxymethyl-2-pyridyl methan-1-ol
[6-(hydroxymethyl)pyridin-2-yl]methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

EINECSNumber : 214-803-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only