missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedimethanol, 98%

Catalog No. aal0252706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0252706 5 g
AAL0252714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0252706 Supplier Thermo Scientific Chemicals Supplier No. L0252706
Only null left

CAS: 1195-59-1 | C7H9NO2 | 139.154 g/mol

2,6-Pyridinedimethanol was used in the preparation of Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) and Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)). It is a ligand used in the synthesis of a variety of metal complexes and catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Pyridinedimethanol was used in the preparation of Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) and Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)). It is a ligand used in the synthesis of a variety of metal complexes and catalysts.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1195-59-1
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00006351
InChI Key WWFMINHWJYHXHF-UHFFFAOYSA-N
Synonym 2,6-pyridinedimethanol, pyridine-2,6-diyldimethanol, 2,6-bis hydroxymethyl pyridine, 6-hydroxymethyl pyridin-2-yl methanol, pyridine-2,6-dimethanol, 2,6-dihydroxymethylpyridine, 2,6-bis-hydroxymethyl pyridine, 2,6-bis-hydroxymethyl-pyridine, 2,6-pyridinyl dimethanol, 6-hydroxymethyl-2-pyridyl methan-1-ol
PubChem CID 70957
IUPAC Name [6-(hydroxymethyl)pyridin-2-yl]methanol
SMILES C1=CC(=NC(=C1)CO)CO

Specifications

Melting Point 110°C to 114°C
Boiling Point 185°C (15 mmHg)
Quantity 5 g
Beilstein 116016
Solubility Information Soluble in water.
Formula Weight 139.15
Percent Purity 98%
Chemical Name or Material 2,6-Pyridinedimethanol
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 214-803-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only