missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedicarboxylic Acid 98.0+%, TCI America™

Catalog No. P055425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P055425G 25 g
P0554100G 100 g
P0554500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P055425G Supplier TCI America Supplier No. P055425G

Chemical Identifiers

CAS 499-83-2
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006299
InChI Key WJJMNDUMQPNECX-UHFFFAOYSA-N
Synonym 2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid
PubChem CID 10367
ChEBI CHEBI:46837
IUPAC Name pyridine-2,6-dicarboxylic acid
SMILES C1=CC(=NC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 252°C
Color White
Quantity 25 g
Formula Weight 167.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Pyridinedicarboxylic Acid
EINECSNumber (5)-0745
TSCA Yes