missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedicarboxylic acid, ≥99.5% (T), MilliporeSigma™ Supelco™

Catalog No. p-7187699
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014333 5 g
111014332 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014333 Supplier Merck Emd Millipore Supplier No. 023215GF
Only null left

Chemical Identifiers

MDL Number MFCD00006299
Synonym DPA; DPAc; Dipicolinic acid

Specifications

Melting Point 248°C to 250°C
CAS 499-83-2
Flash Point 188°C - Closed Cup
Linear Formula C7H5 NO4
Quantity 5 g
Solubility Information Water: 1%, clear
Formula Weight 167.12
Percent Purity ≥99.5% (T)
Grade For ion chromatography