missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Pyridinedicarboxylic acid, ≥99.5% (T), MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | 248°C to 250°C |
| CAS | 499-83-2 |
| Flash Point | 188°C - Closed Cup |
| Linear Formula | C7H5 NO4 |
| Quantity | 10 g |
| Solubility Information | Water: 1%, clear |
| Formula Weight | 167.12 |
| Percent Purity | ≥99.5% (T) |
| Grade | For ion chromatography |