missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Pyridinedicarboxaldehyde 98.0+%, TCI America™
$260.97 - $777.32
Chemical Identifiers
| CAS | 5431-44-7 |
|---|---|
| Molecular Formula | C7H5NO2 |
| Molecular Weight (g/mol) | 135.122 |
| MDL Number | MFCD00010103 |
| InChI Key | PMWXGSWIOOVHEQ-UHFFFAOYSA-N |
| Synonym | 2,6-pyridinedicarboxaldehyde, 2,6-diformylpyridine, 2,6-pyridine dialdehyde, 2,6-pyridinedicarbaldehyde, pubchem22285, acmc-1ap0d, pyda72, 2,6-pyridinedicarboxadehyde, 2,6-pyridine dicarboxaldehyde, 2,6-pyridinedicarbaldehyde # |
| PubChem CID | 79485 |
| IUPAC Name | pyridine-2,6-dicarbaldehyde |
| SMILES | C1=CC(=NC(=C1)C=O)C=O |
Chemical Identifiers
| 5431-44-7 | |
| 135.122 | |
| PMWXGSWIOOVHEQ-UHFFFAOYSA-N | |
| 79485 | |
| C1=CC(=NC(=C1)C=O)C=O |
| C7H5NO2 | |
| MFCD00010103 | |
| 2,6-pyridinedicarboxaldehyde, 2,6-diformylpyridine, 2,6-pyridine dialdehyde, 2,6-pyridinedicarbaldehyde, pubchem22285, acmc-1ap0d, pyda72, 2,6-pyridinedicarboxadehyde, 2,6-pyridine dicarboxaldehyde, 2,6-pyridinedicarbaldehyde # | |
| pyridine-2,6-dicarbaldehyde |
Specifications
| 5431-44-7 | |
| White-Yellow | |
| MFCD00010103 | |
| 2,6-pyridinedicarboxaldehyde, 2,6-diformylpyridine, 2,6-pyridine dialdehyde, 2,6-pyridinedicarbaldehyde, pubchem22285, acmc-1ap0d, pyda72, 2,6-pyridinedicarboxadehyde, 2,6-pyridine dicarboxaldehyde, 2,6-pyridinedicarbaldehyde # | |
| C1=CC(=NC(=C1)C=O)C=O | |
| 135.122 | |
| 135.12 | |
| Crystalline Powder |
| 122°C | |
| C7H5NO2 | |
| 1 g | |
| PMWXGSWIOOVHEQ-UHFFFAOYSA-N | |
| pyridine-2,6-dicarbaldehyde | |
| 79485 | |
| ≥98.0% (GC) | |
| 2,6-Pyridinedicarboxaldehyde |
Safety and Handling
TSCA : No