missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedicarboxaldehyde 98.0+%, TCI America™

Catalog No. P09491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09491G 1 g
P09495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09491G Supplier TCI America Supplier No. P09491G

Chemical Identifiers

CAS 5431-44-7
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.122
MDL Number MFCD00010103
InChI Key PMWXGSWIOOVHEQ-UHFFFAOYSA-N
Synonym 2,6-pyridinedicarboxaldehyde, 2,6-diformylpyridine, 2,6-pyridine dialdehyde, 2,6-pyridinedicarbaldehyde, pubchem22285, acmc-1ap0d, pyda72, 2,6-pyridinedicarboxadehyde, 2,6-pyridine dicarboxaldehyde, 2,6-pyridinedicarbaldehyde #
PubChem CID 79485
IUPAC Name pyridine-2,6-dicarbaldehyde
SMILES C1=CC(=NC(=C1)C=O)C=O

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 1 g
Formula Weight 135.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Pyridinedicarboxaldehyde