missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-(6-Methoxy-2-naphthyl)propionic Acid 98.0+%, TCI America™

Catalog No. M12205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12205G 5 g
M122025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12205G Supplier TCI America Supplier No. M12205G
Only null left

Chemical Identifiers

CAS 23981-80-8
Molecular Formula C14H14O3
Molecular Weight (g/mol) 230.263
MDL Number MFCD00439456
InChI Key CMWTZPSULFXXJA-UHFFFAOYSA-N
Synonym (+/-)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid, (+/-)-Naproxen
PubChem CID 1302
IUPAC Name 2-(6-methoxynaphthalen-2-yl)propanoic acid
SMILES CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O

Specifications

Melting Point 157°C
Quantity 5 g
UN Number 2811
Formula Weight 230.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-2-(6-Methoxy-2-naphthyl)propionic Acid