missing translation for 'onlineSavingsMsg'
Learn More

2,6-Lutidine 98.0+%, TCI America™

Catalog No. p-7131802
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L006725ML 25 mL
L0067500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L006725ML Supplier TCI America Supplier No. L006725ML
Only null left

Chemical Identifiers

CAS 108-48-5
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00006345
InChI Key OISVCGZHLKNMSJ-UHFFFAOYSA-N
Synonym 2,6-lutidine, lutidine, pyridine, 2,6-dimethyl, 2,6-dimethypyridine, alpha,alpha'-lutidine, alpha,alpha'-dimethylpyridine, 2,6-dimethyl-pyridine, hsdb 79, unii-15fq5d0t3p, 2,6-lutidene
PubChem CID 7937
ChEBI CHEBI:32548
IUPAC Name 2,6-dimethylpyridine
SMILES CC1=CC=CC(C)=N1

Specifications

Melting Point -6°C
Color Yellow
Boiling Point 144°C
Quantity 25 mL
UN Number 1993
Formula Weight 107.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Lutidine
EINECSNumber (5)-0712
RTECSNumber OK9700000
TSCA Yes