missing translation for 'onlineSavingsMsg'
Learn More

2,6-Lutidine N-Oxide 98.0+%, TCI America™

Catalog No. p-7131804
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00695G 5 g
L006925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L00695G Supplier TCI America Supplier No. L00695G
Only null left

Chemical Identifiers

CAS 1073-23-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00039715
InChI Key LIDGFHXPUOJZMK-UHFFFAOYSA-N
Synonym 2,6-dimethylpyridine n-oxide, 2,6-lutidine n-oxide, 2,6-lutidine-n-oxide, 2,6-dimethylpyridine 1-oxide, 2,6-lutidine 1-oxide, pyridine, 2,6-dimethyl-, 1-oxide, 2,6-lutidine oxide, 2,6-dimethylpyridine oxide, 2,6-dimethylpyridinium n-oxide, 2,6-dimethylpyridin-1-ium-1-olate
PubChem CID 14083
IUPAC Name 2,6-dimethyl-1-oxidopyridin-1-ium
SMILES CC1=[N+](C(=CC=C1)C)[O-]

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Lutidine N-Oxide