missing translation for 'onlineSavingsMsg'
Learn More

2,6-Lutidine N-oxide, 98%

CAS: 1073-23-0 | C7H9NO | 123.155 g/mol

Supplier:  Thermo Scientific Chemicals A1429909

 View more versions of this product

Catalog No. AAA1429909


Only null left
Explore available promotions

Description

Description

2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents, moisture and water.
Specifications

Specifications

2,6-Lutidine N-oxide
1.102
C7H9NO
MFCD00039715
110551
2,6-dimethylpyridine n-oxide, 2,6-lutidine n-oxide, 2,6-lutidine-n-oxide, 2,6-dimethylpyridine 1-oxide, 2,6-lutidine 1-oxide, pyridine, 2,6-dimethyl-, 1-oxide, 2,6-lutidine oxide, 2,6-dimethylpyridine oxide, 2,6-dimethylpyridinium n-oxide, 2,6-dimethylpyridin-1-ium-1-olate
LIDGFHXPUOJZMK-UHFFFAOYSA-N
2,6-dimethyl-1-oxidopyridin-1-ium
14083
98%
1073-23-0
108°C to 111°C (10 mmHg)
1.5685
10 g
Hygroscopic
Miscible with water.
CC1=[N+](C(=CC=C1)C)[O-]
123.155
123.16

Chemical Identifiers

1073-23-0
123.155
LIDGFHXPUOJZMK-UHFFFAOYSA-N
14083
CC1=[N+](C(=CC=C1)C)[O-]
C7H9NO
MFCD00039715
2,6-dimethylpyridine n-oxide, 2,6-lutidine n-oxide, 2,6-lutidine-n-oxide, 2,6-dimethylpyridine 1-oxide, 2,6-lutidine 1-oxide, pyridine, 2,6-dimethyl-, 1-oxide, 2,6-lutidine oxide, 2,6-dimethylpyridine oxide, 2,6-dimethylpyridinium n-oxide, 2,6-dimethylpyridin-1-ium-1-olate
2,6-dimethyl-1-oxidopyridin-1-ium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

EINECSNumber : 214-025-1

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only