missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2,6-Lutidine

Catalog No. 3361062L
Encompass
Change view
Click to view available options
Quantity:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3361062L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3361062L Supplier Sigma Aldrich Supplier No. 3361062L

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,6-Lutidine

Specifications

CAS 108-48-5
Melting Point -6°C
Density 0.92 g/mL at 25°C
Boiling Point 143°C to 145°C
Molecular Formula C7H9N
Refractive Index n20/D 1.497
Linear Formula C7H9N
MDL Number MFCD00006345
Quantity 2 L
Synonym 2,6-Dimethylpyridine
Molecular Weight (g/mol) 107.15 g/mol
Show More Show Less