missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diphenylpyridine 99.0+%, TCI America™

Catalog No. p-7127015
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19225G 5 g
D192225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19225G Supplier TCI America Supplier No. D19225G
Only null left

Chemical Identifiers

CAS 3558-69-8
Molecular Formula C17H13N
Molecular Weight (g/mol) 231.298
MDL Number MFCD00006284
InChI Key PJUOHDQXFNPPRF-UHFFFAOYSA-N
Synonym pyridine, 2,6-diphenyl, unii-zi75v15y1t, 2,6-diphenyl-pyridine, 2,6-diphenyl pyridine, maybridge3_000511, acmc-1cq41, pjuohdqxfnpprf-uhfffaoysa
PubChem CID 72920
IUPAC Name 2,6-diphenylpyridine
SMILES C1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=CC=C3

Specifications

Color White-Yellow
Boiling Point 398°C
Quantity 5 g
Formula Weight 231.30
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diphenylpyridine