missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diphenylpyridine, 97%

Catalog No. p-7028058
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2517906 5 g
AAB2517914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2517906 Supplier Thermo Scientific Chemicals Supplier No. B2517906
Only null left

CAS: 3558-69-8 | C17H13N | 231.298 g/mol

2,6-Diphenylpyridine is used as a tridentate [C?N?C] dianionic ligand in organogold(III) chemistry, an intermediate. It readily adds an electron from alkali metals in THF to give a radical anion which is a powerful reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diphenylpyridine is used as a tridentate [C∧N∧C] dianionic ligand in organogold(III) chemistry, an intermediate. It readily adds an electron from alkali metals in THF to give a radical anion which is a powerful reducing agent.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3558-69-8
Molecular Formula C17H13N
Molecular Weight (g/mol) 231.298
MDL Number MFCD00006284
InChI Key PJUOHDQXFNPPRF-UHFFFAOYSA-N
Synonym pyridine, 2,6-diphenyl, unii-zi75v15y1t, 2,6-diphenyl-pyridine, 2,6-diphenyl pyridine, maybridge3_000511, acmc-1cq41, pjuohdqxfnpprf-uhfffaoysa
PubChem CID 72920
IUPAC Name 2,6-diphenylpyridine
SMILES C1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 74°C to 81°C
Boiling Point 210°C (3 mmHg)
Quantity 5 g
Beilstein 7516
Solubility Information Insoluble in water.
Formula Weight 231.3
Percent Purity 97%
Chemical Name or Material 2,6-Diphenylpyridine
EINECSNumber 222-620-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only