missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylquinoline 99.0+%, TCI America™
$67.11 - $596.49
Chemical Identifiers
| CAS | 877-43-0 |
|---|---|
| Molecular Formula | C11H11N |
| Molecular Weight (g/mol) | 157.216 |
| MDL Number | MFCD00006762 |
| InChI Key | JJPSZKIOGBRMHK-UHFFFAOYSA-N |
| Synonym | p-toluquinaldine, quinoline, 2,6-dimethyl, 6-methylquinaldine, unii-kst0m1t4mb, 2,6-dimethyl-quinoline, kst0m1t4mb, pubchem5876, quinoline,6-dimethyl, acmc-209qpt, 2,6-dimethylquinoline; |
| PubChem CID | 13414 |
| IUPAC Name | 2,6-dimethylquinoline |
| SMILES | CC1=CC2=C(C=C1)N=C(C=C2)C |
Chemical Identifiers
| 877-43-0 | |
| 157.216 | |
| JJPSZKIOGBRMHK-UHFFFAOYSA-N | |
| 13414 | |
| CC1=CC2=C(C=C1)N=C(C=C2)C |
| C11H11N | |
| MFCD00006762 | |
| p-toluquinaldine, quinoline, 2,6-dimethyl, 6-methylquinaldine, unii-kst0m1t4mb, 2,6-dimethyl-quinoline, kst0m1t4mb, pubchem5876, quinoline,6-dimethyl, acmc-209qpt, 2,6-dimethylquinoline; | |
| 2,6-dimethylquinoline |
Specifications
| 877-43-0 | |
| White | |
| C11H11N | |
| 1 g | |
| JJPSZKIOGBRMHK-UHFFFAOYSA-N | |
| 2,6-dimethylquinoline | |
| 13414 | |
| ≥99.0% (GC) | |
| 2,6-Dimethylquinoline |
| 58°C | |
| 267°C | |
| MFCD00006762 | |
| p-toluquinaldine, quinoline, 2,6-dimethyl, 6-methylquinaldine, unii-kst0m1t4mb, 2,6-dimethyl-quinoline, kst0m1t4mb, pubchem5876, quinoline,6-dimethyl, acmc-209qpt, 2,6-dimethylquinoline; | |
| CC1=CC2=C(C=C1)N=C(C=C2)C | |
| 157.216 | |
| 157.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes