missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylquinoline, 98%

CAS: 877-43-0 | C11H11N | 157.216 g/mol

Supplier:  Thermo Scientific Chemicals B2393414

 View more versions of this product

Catalog No. AAB2393414


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,6-Dimethylquinoline
56°C to 58°C
C11H11N
25 g
JJPSZKIOGBRMHK-UHFFFAOYSA-N
2,6-dimethylquinoline
13414
98%
877-43-0
266°C to 267°C
MFCD00006762
p-toluquinaldine, quinoline, 2,6-dimethyl, 6-methylquinaldine, unii-kst0m1t4mb, 2,6-dimethyl-quinoline, kst0m1t4mb, pubchem5876, quinoline,6-dimethyl, acmc-209qpt, 2,6-dimethylquinoline;
CC1=CC2=C(C=C1)N=C(C=C2)C
157.216
157.21

Chemical Identifiers

877-43-0
157.216
JJPSZKIOGBRMHK-UHFFFAOYSA-N
13414
CC1=CC2=C(C=C1)N=C(C=C2)C
C11H11N
MFCD00006762
p-toluquinaldine, quinoline, 2,6-dimethyl, 6-methylquinaldine, unii-kst0m1t4mb, 2,6-dimethyl-quinoline, kst0m1t4mb, pubchem5876, quinoline,6-dimethyl, acmc-209qpt, 2,6-dimethylquinoline;
2,6-dimethylquinoline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 212-891-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only