missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylpiperazine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 116671000
Description
Specifications
| 2,6-Dimethylpiperazine | |
| 162.0°C | |
| Authentic | |
| C6H14N2 | |
| 100g | |
| piperazine, 2,6-dimethyl, 3,5-dimethylpiperazine, 2,6-dimethyl-piperazine, 2,6-dimethyl piperazine, 2r,6s-rel-2,6-dimethylpiperazine, pubchem8571, 2,6 dimethylpiperazine, 2,6-dimehtylpiperazine, piperazine,6-dimethyl, acmc-1bolo | |
| IFNWESYYDINUHV-UHFFFAOYNA-N | |
| 2,6-dimethylpiperazine | |
| 66056 | |
| 98% |
| 108-49-6 | |
| 44°C | |
| 97.5% min. (GC) | |
| MFCD07772435,MFCD00005956 | |
| 23,I,8 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in most organic solvents | |
| CC1CNCC(C)N1 | |
| 114.19 | |
| 114.19 |
Chemical Identifiers
| 108-49-6 | |
| 114.19 | |
| IFNWESYYDINUHV-UHFFFAOYNA-N | |
| 66056 | |
| CC1CNCC(C)N1 |
| C6H14N2 | |
| MFCD07772435,MFCD00005956 | |
| piperazine, 2,6-dimethyl, 3,5-dimethylpiperazine, 2,6-dimethyl-piperazine, 2,6-dimethyl piperazine, 2r,6s-rel-2,6-dimethylpiperazine, pubchem8571, 2,6 dimethylpiperazine, 2,6-dimehtylpiperazine, piperazine,6-dimethyl, acmc-1bolo | |
| 2,6-dimethylpiperazine |
Safety and Handling
GHS H Statement
Flammable solid.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 203-588-9
RUO â Research Use Only