missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylpiperazine, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 108-49-6 |
|---|---|
| Molecular Formula | C6H14N2 |
| Molecular Weight (g/mol) | 114.19 |
| MDL Number | MFCD07772435,MFCD00005956 |
| InChI Key | IFNWESYYDINUHV-UHFFFAOYNA-N |
| Synonym | piperazine, 2,6-dimethyl, 3,5-dimethylpiperazine, 2,6-dimethyl-piperazine, 2,6-dimethyl piperazine, 2r,6s-rel-2,6-dimethylpiperazine, pubchem8571, 2,6 dimethylpiperazine, 2,6-dimehtylpiperazine, piperazine,6-dimethyl, acmc-1bolo |
| PubChem CID | 66056 |
| IUPAC Name | 2,6-dimethylpiperazine |
| SMILES | CC1CNCC(C)N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC116670250
|
N/A | 25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C6H14N2 | |
| MFCD07772435,MFCD00005956 | |
| piperazine, 2,6-dimethyl, 3,5-dimethylpiperazine, 2,6-dimethyl-piperazine, 2,6-dimethyl piperazine, 2r,6s-rel-2,6-dimethylpiperazine, pubchem8571, 2,6 dimethylpiperazine, 2,6-dimehtylpiperazine, piperazine,6-dimethyl, acmc-1bolo | |
| 2,6-dimethylpiperazine |
Specifications
| 108-49-6 | |
| 44°C | |
| C6H14N2 | |
| 23,I,8 | |
| Solubility in water: slightly soluble. Other solubilities: soluble in most organic solvents | |
| CC1CNCC(C)N1 | |
| 114.19 | |
| 114.19 | |
| 2, 6-Dimethylpiperazine |
| 162.0°C | |
| 97.5% min. (GC) | |
| MFCD07772435,MFCD00005956 | |
| piperazine, 2,6-dimethyl, 3,5-dimethylpiperazine, 2,6-dimethyl-piperazine, 2,6-dimethyl piperazine, 2r,6s-rel-2,6-dimethylpiperazine, pubchem8571, 2,6 dimethylpiperazine, 2,6-dimehtylpiperazine, piperazine,6-dimethyl, acmc-1bolo | |
| IFNWESYYDINUHV-UHFFFAOYNA-N | |
| 2,6-dimethylpiperazine | |
| 66056 | |
| 98% |
Safety and Handling
GHS H Statement
Flammable solid.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 203-588-9
RUO – Research Use Only