missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylbenzoic acid, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 169010050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC169010050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

632-46-2
150.18
HCBHQDKBSKYGCK-UHFFFAOYSA-N
12439
2,6-dimethylbenzoic acid
C9H10O2
MFCD00002483
benzoic acid, 2,6-dimethyl, 2,6-dimethylbenzoicacid, 2,6-dimethyl-benzoic acid, vic-m-xylylic acid, 2,6-dimethyl benzoic acid, m-xylene-2-carboxylic acid, 2,6-dimethylbenzene carboxylic acid, benzoic acid, 2,6-dimethyl-7ci,8ci,9ci, zlchem 286, vic.-m-xylylic acid
CHEBI:64827
CC1=C(C(=CC=C1)C)C(=O)O

Specifications

2, 6-Dimethylbenzoic acid
98.5
Authentic
Glass bottle
(CH3)2C6H3COOH
MFCD00002483
benzoic acid, 2,6-dimethyl, 2,6-dimethylbenzoicacid, 2,6-dimethyl-benzoic acid, vic-m-xylylic acid, 2,6-dimethyl benzoic acid, m-xylene-2-carboxylic acid, 2,6-dimethylbenzene carboxylic acid, benzoic acid, 2,6-dimethyl-7ci,8ci,9ci, zlchem 286, vic.-m-xylylic acid
CC1=C(C(=CC=C1)C)C(=O)O
150.18
CHEBI:64827
99%
632-46-2
100.0
99%
C9H10O2
5g
01, 259
HCBHQDKBSKYGCK-UHFFFAOYSA-N
2,6-dimethylbenzoic acid
12439
150.18
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-177-

SDS
Documents

Documents