missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylanisole 98.0+%, TCI America™

Catalog No. d238325ml
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D238325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D238325ML Supplier TCI America Supplier No. D238325ML
Only null left

Chemical Identifiers

CAS 1004-66-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00008380
InChI Key GFNZJAUVJCGWLW-UHFFFAOYSA-N
Synonym 2,6-dimethylanisole, 2-methoxy-m-xylene, benzene, 2-methoxy-1,3-dimethyl, 1,3-dimethyl-2-methoxybenzene, pubchem4115, 2,6-dimethyl anisole, acmc-1buuc, ksc493q8r, 2,6-dimethylphenyl methyl ether
PubChem CID 66088
IUPAC Name 2-methoxy-1,3-dimethylbenzene
SMILES COC1=C(C)C=CC=C1C

Specifications

Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,6-Dimethylanisole