missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethyl-gamma-pyrone 99.0+%, TCI America™
$157.87 - $411.98
Chemical Identifiers
| CAS | 1004-36-0 |
|---|---|
| Molecular Formula | C7H8O2 |
| Molecular Weight (g/mol) | 124.139 |
| MDL Number | MFCD00006579 |
| InChI Key | VSYFZULSKMFUJJ-UHFFFAOYSA-N |
| Synonym | 2,6-dimethyl-4h-pyran-4-one, 2,6-dimethyl-4-pyrone, 2,6-dimethyl-gamma-pyrone, 2,6-dimethyl-4-pyranone, 4h-pyran-4-one, 2,6-dimethyl, gamma-2,6-dimethylpyrone, unii-v2d262mf0l, 2,6-dimethyl-pyran-4-one, 2,6-dimethyl-.gamma.-pyrone, ccris 3601 |
| PubChem CID | 13862 |
| IUPAC Name | 2,6-dimethylpyran-4-one |
| SMILES | CC1=CC(=O)C=C(O1)C |
Chemical Identifiers
| 1004-36-0 | |
| 124.139 | |
| VSYFZULSKMFUJJ-UHFFFAOYSA-N | |
| 13862 | |
| CC1=CC(=O)C=C(O1)C |
| C7H8O2 | |
| MFCD00006579 | |
| 2,6-dimethyl-4h-pyran-4-one, 2,6-dimethyl-4-pyrone, 2,6-dimethyl-gamma-pyrone, 2,6-dimethyl-4-pyranone, 4h-pyran-4-one, 2,6-dimethyl, gamma-2,6-dimethylpyrone, unii-v2d262mf0l, 2,6-dimethyl-pyran-4-one, 2,6-dimethyl-.gamma.-pyrone, ccris 3601 | |
| 2,6-dimethylpyran-4-one |
Specifications
| 1004-36-0 | |
| White-Yellow | |
| MFCD00006579 | |
| 2,6-dimethyl-4h-pyran-4-one, 2,6-dimethyl-4-pyrone, 2,6-dimethyl-gamma-pyrone, 2,6-dimethyl-4-pyranone, 4h-pyran-4-one, 2,6-dimethyl, gamma-2,6-dimethylpyrone, unii-v2d262mf0l, 2,6-dimethyl-pyran-4-one, 2,6-dimethyl-.gamma.-pyrone, ccris 3601 | |
| CC1=CC(=O)C=C(O1)C | |
| 124.139 | |
| 124.14 | |
| Crystalline Powder |
| 134°C | |
| C7H8O2 | |
| 5 g | |
| VSYFZULSKMFUJJ-UHFFFAOYSA-N | |
| 2,6-dimethylpyran-4-one | |
| 13862 | |
| ≥99.0% (GC) | |
| 2,6-Dimethyl-gamma-pyrone |
Safety and Handling
EINECSNumber : (5)-0251
RTECSNumber : UQ0780000
TSCA : No