missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-gamma-pyrone 99.0+%, TCI America™

Catalog No. D07945G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
D07945G 5 g
D079425G 25 g
2 options

Catalog No. D07945G

Supplier: TCI America D07945G

Only null left

Chemical Identifiers

CAS 1004-36-0
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00006579
InChI Key VSYFZULSKMFUJJ-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4h-pyran-4-one, 2,6-dimethyl-4-pyrone, 2,6-dimethyl-gamma-pyrone, 2,6-dimethyl-4-pyranone, 4h-pyran-4-one, 2,6-dimethyl, gamma-2,6-dimethylpyrone, unii-v2d262mf0l, 2,6-dimethyl-pyran-4-one, 2,6-dimethyl-.gamma.-pyrone, ccris 3601
PubChem CID 13862
IUPAC Name 2,6-dimethylpyran-4-one
SMILES CC1=CC(=O)C=C(O1)C

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 5 g
Formula Weight 124.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethyl-gamma-pyrone
EINECSNumber (5)-0251
RTECSNumber UQ0780000
TSCA No