missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-heptanone, >90% (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)

Catalog No. AAL160150F
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL160150F 2500 mL
AAL16015AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL160150F Supplier Thermo Scientific Chemicals Supplier No. L160150F
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-83-8 | C9H18O | 142.242 g/mol

2,6-Dimethyl-4-heptanone is used as a coating solvent. It is an active component of mint oil. It acts as a dispersant for organosol type resins. It is involved in the antigerminative treatment of bulbs and tubers. Further, it is used as a solvent for nitrocellulose. In addition to this, it acts as an intermediate in the preparation of inhibitors, active pharmaceutical ingredients and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethyl-4-heptanone is used as a coating solvent. It is an active component of mint oil. It acts as a dispersant for organosol type resins. It is involved in the antigerminative treatment of bulbs and tubers. Further, it is used as a solvent for nitrocellulose. In addition to this, it acts as an intermediate in the preparation of inhibitors, active pharmaceutical ingredients and dyes.

Solubility
Miscible with ethanol, ether, carbon tetrachloride, chloroform, benzene and most organic liquids.

Notes
Incompatible with strong bases, strong oxidizing agents, strong acids and strong reducing agents.

Chemical Identifiers

CAS 108-83-8
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00008940
InChI Key PTTPXKJBFFKCEK-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-heptanone, diisobutyl ketone, isovalerone, isobutyl ketone, valerone, diisobutylketone, diisobutilchetone, s-diisopropylacetone, di-isobutylcetone, dibk
PubChem CID 7958
IUPAC Name 2,6-dimethylheptan-4-one
SMILES CC(C)CC(=O)CC(C)C

Specifications

Melting Point -42°C
Density 0.809
Boiling Point 163°C to 173°C
Flash Point 47°C (117°F)
Odor Unpleasant
Refractive Index 1.414
Quantity 2500 mL
UN Number UN1224
Beilstein 1743163
Solubility Information Miscible with ethanol,ether,carbon tetrachloride,chloroform,benzene and most organic liquids.
Formula Weight 142.24
Percent Purity >90%
Chemical Name or Material 2,6-Dimethyl-4-heptanone, (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)
Show More Show Less
Hazard Category H226-H335
Hazard Statement GHS H Statement
H226-H335
Flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 203-620-1
RTECSNumber MJ5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only