missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-heptanol 85.0+%, TCI America™

Catalog No. D073225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D073225ML 25 mL
D0732500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D073225ML Supplier TCI America Supplier No. D073225ML
Only null left

Chemical Identifiers

CAS 108-82-7
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
MDL Number MFCD00008944
InChI Key HXQPUEQDBSPXTE-UHFFFAOYSA-N
Synonym Diisobutylcarbinol
PubChem CID 7957
IUPAC Name 2,6-dimethylheptan-4-ol
SMILES CC(C)CC(CC(C)C)O

Specifications

Color Yellow
Boiling Point 179°C
Quantity 25 mL
Formula Weight 144.26
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dimethyl-4-heptanol
EINECSNumber (2)-0217
RTECSNumber MJ3325000
TSCA Yes