missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-1,4-benzoquinone 98.0+%, TCI America™

Catalog No. D22345G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22345G 5 g
D22341G 1 g
D223425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D22345G Supplier TCI America Supplier No. D22345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 527-61-7
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00001605
InChI Key SENUUPBBLQWHMF-UHFFFAOYSA-N
Synonym 2,6-dimethylbenzoquinone, 2,6-dimethyl-1,4-benzoquinone, 2,6-dimethyl-p-benzoquinone, m-xyloquinone, 2,6-xyloquinone, 2,6-dimethylquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-dimethyl, 3,5-dimethylbenzoquinone, p-benzoquinone, 2,6-dimethyl, 2,6-dimethyl-1,4-quinone
PubChem CID 68241
IUPAC Name 2,6-dimethylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=CC(=O)C=C(C1=O)C

Specifications

Melting Point 73°C
Quantity 5 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethyl-1,4-benzoquinone
RTECSNumber DK4825000
TSCA No