missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethoxypyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
$160.30 - $160.30
Specifications
| CAS | 221006-70-8 |
|---|---|
| Melting Point | 109°C |
| Color | White |
| Molecular Formula | C7H10BNO4 |
| MDL Number | MFCD03788239 |
Specifications
| 221006-70-8 | |
| White | |
| MFCD03788239 | |
| B(C1=C(N=C(C=C1)OC)OC)(O)O | |
| 182.97 | |
| 182.97 |
| 109°C | |
| C7H10BNO4 | |
| ADGHSWFUZUADDH-UHFFFAOYSA-N | |
| (2,6-dimethoxypyridin-3-yl)boronic acid | |
| 2762707 | |
| Crystalline Powder |