missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxypyridine-3-boronic acid, 95%

Catalog No. AAL2038903
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2038903 1 g
AAL20389MD 250 mg
AAL2038906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL2038903 Supplier Thermo Scientific Chemicals Supplier No. L2038903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 221006-70-8 | C7H10BNO4 | 182.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 221006-70-8
Molecular Formula C7H10BNO4
Molecular Weight (g/mol) 182.97
MDL Number MFCD03788239
InChI Key ADGHSWFUZUADDH-UHFFFAOYSA-N
Synonym 2,6-dimethoxypyridine-3-boronic acid, 2,6-dimethoxy-3-pyridineboronic acid, 2,6-dimethoxypyridin-3-yl boronic acid, 2,6-dimethoxypyridin-3-yl-3-boronic acid, 3-borono-2,6-dimethoxypyridine, 2,6-dimethoxy-3-pyridylboronic acid, 2,6-dimethoxypyridine-5-boronic acid, boronic acid, 2,6-dimethoxy-3-pyridinyl, 2,6-dimethoxy-3-pyridyl boronic acid
PubChem CID 2762707
IUPAC Name (2,6-dimethoxypyridin-3-yl)boronic acid
SMILES B(C1=C(N=C(C=C1)OC)OC)(O)O

Specifications

Melting Point 128°C to 130°C
Quantity 1 g
Beilstein 9391039
Formula Weight 182.97
Percent Purity 95%
Chemical Name or Material 2,6-Dimethoxypyridine-3-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only