missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxybenzaldehyde 97.0+%, TCI America™

Catalog No. p-7127494
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27785G 5 g
D277825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27785G Supplier TCI America Supplier No. D27785G

Chemical Identifiers

CAS 3392-97-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00010862
InChI Key WXSGQHKHUYTJNB-UHFFFAOYSA-N
Synonym benzaldehyde, 2,6-dimethoxy, unii-54p7cg8v99, pubchem8226, acmc-1ckys, 2,6 dimethoxybenzaldehyde, 2,6-dimethoxy-benzaldehyde, ksc497m3h, 1,3-dimethoxy-2-formylbenzene, paragos 460025, timtec-bb sbb006541
PubChem CID 96404
IUPAC Name 2,6-dimethoxybenzaldehyde
SMILES COC1=CC=CC(OC)=C1C=O

Specifications

Melting Point 97°C
Color White-Yellow
Boiling Point 158°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethoxybenzaldehyde
EINECSNumber (3)-2665
TSCA No