missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxybenzaldehyde, 98+%

Catalog No. p-7081933
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1220903 1 g
AAA1220906 5 g
AAA1220914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1220903 Supplier Thermo Scientific Chemicals Supplier No. A1220903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3392-97-0 | C9H10O3 | 166.18 g/mol

2,6-Dimethoxybenzaldehyde has been used in the preparation of 2,6-dihydroxybenzaldehyde by demethylation with AlBr3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethoxybenzaldehyde has been used in the preparation of 2,6-dihydroxybenzaldehyde by demethylation with AlBr3.

Solubility
Soluble in Methanol (almost transparency).

Notes
Air sensitive. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent, air.

Chemical Identifiers

CAS 3392-97-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00010862
InChI Key WXSGQHKHUYTJNB-UHFFFAOYSA-N
Synonym benzaldehyde, 2,6-dimethoxy, unii-54p7cg8v99, pubchem8226, acmc-1ckys, 2,6 dimethoxybenzaldehyde, 2,6-dimethoxy-benzaldehyde, ksc497m3h, 1,3-dimethoxy-2-formylbenzene, paragos 460025, timtec-bb sbb006541
PubChem CID 96404
IUPAC Name 2,6-dimethoxybenzaldehyde
SMILES COC1=CC=CC(OC)=C1C=O

Specifications

Melting Point 96°C to 98°C
Boiling Point 285°C
Quantity 1 g
Beilstein 908138
Sensitivity Air Sensitive
Solubility Information Soluble in Methanol (almost transparency).
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material 2,6-Dimethoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only