missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',6'-Dimethoxyacetophenone 98.0+%, TCI America™
$32.72 - $84.70
Chemical Identifiers
| CAS | 2040-04-2 |
|---|---|
| Molecular Formula | C10H12O3 |
| Molecular Weight (g/mol) | 180.20 |
| MDL Number | MFCD00008729 |
| InChI Key | XEUGKOFTNAYMMX-UHFFFAOYSA-N |
| Synonym | 2',6'-dimethoxyacetophenone, 1-2,6-dimethoxyphenyl ethanone, 2,6-dimethoxyacetophenone, acetophenone, 2',6'-dimethoxy, 1-2,6-dimethoxyphenyl ethan-1-one, ethanone, 1-2,6-dimethoxyphenyl, usaf k-2801, 2,6-dimethoxy acetophenone, 1-acetyl-2,6-dimethoxybenzene, pubchem3380 |
| PubChem CID | 16267 |
| IUPAC Name | 1-(2,6-dimethoxyphenyl)ethanone |
| SMILES | CC(=O)C1=C(C=CC=C1OC)OC |
Chemical Identifiers
| 2040-04-2 | |
| 180.20 | |
| XEUGKOFTNAYMMX-UHFFFAOYSA-N | |
| 16267 | |
| CC(=O)C1=C(C=CC=C1OC)OC |
| C10H12O3 | |
| MFCD00008729 | |
| 2',6'-dimethoxyacetophenone, 1-2,6-dimethoxyphenyl ethanone, 2,6-dimethoxyacetophenone, acetophenone, 2',6'-dimethoxy, 1-2,6-dimethoxyphenyl ethan-1-one, ethanone, 1-2,6-dimethoxyphenyl, usaf k-2801, 2,6-dimethoxy acetophenone, 1-acetyl-2,6-dimethoxybenzene, pubchem3380 | |
| 1-(2,6-dimethoxyphenyl)ethanone |
Specifications
| 2040-04-2 | |
| White | |
| C10H12O3 | |
| 1 g | |
| XEUGKOFTNAYMMX-UHFFFAOYSA-N | |
| 1-(2,6-dimethoxyphenyl)ethanone | |
| 16267 | |
| ≥98.0% (GC) | |
| 2′,6′-Dimethoxyacetophenone |
| 70°C | |
| 136°C | |
| MFCD00008729 | |
| 2',6'-dimethoxyacetophenone, 1-2,6-dimethoxyphenyl ethanone, 2,6-dimethoxyacetophenone, acetophenone, 2',6'-dimethoxy, 1-2,6-dimethoxyphenyl ethan-1-one, ethanone, 1-2,6-dimethoxyphenyl, usaf k-2801, 2,6-dimethoxy acetophenone, 1-acetyl-2,6-dimethoxybenzene, pubchem3380 | |
| CC(=O)C1=C(C=CC=C1OC)OC | |
| 180.20 | |
| 180.20 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : AM8400000
TSCA : No