missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dimethoxyacetophenone 98.0+%, TCI America™

Catalog No. D48491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48491G 1 g
D48495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48491G Supplier TCI America Supplier No. D48491G

Chemical Identifiers

CAS 2040-04-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00008729
InChI Key XEUGKOFTNAYMMX-UHFFFAOYSA-N
Synonym 2',6'-dimethoxyacetophenone, 1-2,6-dimethoxyphenyl ethanone, 2,6-dimethoxyacetophenone, acetophenone, 2',6'-dimethoxy, 1-2,6-dimethoxyphenyl ethan-1-one, ethanone, 1-2,6-dimethoxyphenyl, usaf k-2801, 2,6-dimethoxy acetophenone, 1-acetyl-2,6-dimethoxybenzene, pubchem3380
PubChem CID 16267
IUPAC Name 1-(2,6-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC=C1OC)OC

Specifications

Melting Point 70°C
Color White
Boiling Point 136°C
Quantity 1 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,6′-Dimethoxyacetophenone
RTECSNumber AM8400000
TSCA No