missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dimethoxyacetophenone, 98%

CAS: 2040-04-2 | C8H6Cl2O | 189.04 g/mol

$117.40 - $395.42

Chemical Identifiers

CAS 2040-04-2
MDL Number MFCD00008729
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1673306
View Documents
Thermo Scientific Chemicals
A1673306
5 g
Each for $117.40
Only null left
 
AAA1673314
View Documents
Thermo Scientific Chemicals
A1673314
25 g
Each for $395.42
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2040-04-2
MFCD00008729

Specifications

2040-04-2
135°C to 136°C (2 mmHg)
MFCD00008729
2048976
HYBDSXBLGCQKRE-UHFFFAOYSA-N
1-(2,6-dichlorophenyl)ethanone
74877
98%
68°C to 71°C
C8H6Cl2O
5 g
2',6'-dichloroacetophenone, 1-2,6-dichlorophenyl ethanone, 2,6-dichloroacetophenone, 1-2,6-dichlorophenyl ethan-1-one, ethanone, 1-2,6-dichlorophenyl, 2,6-dichloro acetophenone, 1-acetyl-2,6-dichlorobenzene, zlchem 1084, pubchem12362, acmc-1cobf
CC(=O)C1=C(C=CC=C1Cl)Cl
189.04
180.2
2',6'-Dimethoxyacetophenone
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 218-034-1

RTECSNumber : AM8400000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only